Anti-MCTS1 Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:200 to 1:500
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77844-761
- 77844-762
- Antigen Symbol:
- MCTS1
- MCTS1
- Antigen Synonyms:
- FLJ39637|Malignant T cell amplified sequence 1|Malignant T cell-amplified sequence 1|MCT 1|MCT-1|MCT1|MCTS 1|MCTS1|MCTS1_HUMAN|Multiple copies T cell malignancies 1|Multiple copies T cell malignancies|Multiple copies T-cell malignancies|Oncogene MCT 1|Oncogene MCT1
- FLJ39637|Malignant T cell amplified sequence 1|Malignant T cell-amplified sequence 1|MCT 1|MCT-1|MCT1|MCTS 1|MCTS1|MCTS1_HUMAN|Multiple copies T cell malignancies 1|Multiple copies T cell malignancies|Multiple copies T-cell malignancies|Oncogene MCT 1|Oncogene MCT1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9ULC4
- Q9ULC4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Recombinant Human Malignant T-cell-amplified sequence 1 protein (1-181AA)
- Recombinant Human Malignant T-cell-amplified sequence 1 protein (1-181AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together