Anti-FUT8 Rabbit Recombinant Polyclonal Antibody
Catalog # 77842-976
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:FUT8
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q9BYC5
- Antigen Synonyms:6-fucosyltransferase|Alpha (1,6) fucosyltransferase|Alpha-(1,6)-fucosyltransferase|Alpha1 6FucT|alpha1-6FucT|Fucosyltransferase 8 (alpha (1,6) fucosyltransferase)|Fucosyltransferase 8|FUT8_HUMAN|GDP fucose glycoprotein fucosyltransferase|GDP L Fuc:N acetyl beta D glucosaminide alpha1,6 fucosyltransferase|GDP-fucose--glycoprotein fucosyltransferase|GDP-L-Fuc:N-acetyl-beta-D-glucosaminide alpha1|Glycoprotein 6 alpha L fucosyltransferase|Glycoprotein 6-alpha-L-fucosyltransferase|MGC26465
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Alpha-(1,6)-fucosyltransferase protein (247-377AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:200 to 1:500
Type:
Antigen: FUT8
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse