Anti-TIMM10B Rabbit Recombinant Polyclonal Antibody
Catalog # 77845-442
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:TIMM10B
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q9Y5J6
- Antigen Synonyms:Fracture callus 1 homolog (rat)|Fracture callus 1 homolog|Fracture callus protein 1|FxC1|Mitochondrial import inner membrane translocase subunit Tim10 B|Mitochondrial import inner membrane translocase subunit Tim9 B|OTTHUMP00000164584|OTTHUMP00000230720|TIM 10B|Tim10b|Tim9b|TIM9B_HUMAN|Translocase of inner mitochondrial membrane 10 homolog B (yeast)
- Storage Buffer:0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Mitochondrial import inner membrane translocase subunit Tim10 B protein (1-103AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: TIMM10B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse