Anti-RHOA Rabbit Recombinant Polyclonal Antibody
Catalog # 77855-503
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:RHOA
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P61586
- Antigen Synonyms:ARH12|ARHA|H12|ras homolog gene family member A|ras homolog gene family member B|ras homolog gene family member C|Rho cDNA clone 12|RHO12|RHOA_HUMAN|rhob|rhoc|RHOH12|Small GTP binding protein Transforming protein RhoA
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Transforming protein RhoA protein (1-191AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC:1:20 to 1:200
Type:
Antigen: RHOA
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human