Anti-MC1R Rabbit Recombinant Polyclonal Antibody
Catalog # 77859-550
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:MC1R
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q01726
- Antigen Synonyms:CMM5|MC1 Receptor|MC1-R|MC1R|Melanocortin 1 receptor|Melanocortin 1 receptor (alpha melanocyte stimulating hormone receptor)|Melanocortin receptor 1|Melanocyte-stimulating hormone receptor|Melanotropin receptor|MSH-R|MSHR|MSHR_HUMAN|SHEP2
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Melanocyte-stimulating hormone receptor protein (1-37AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:200 to 1:500, IF: 1:50 to 1:200
Type:
Antigen: MC1R
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human