Anti-CDKN1A Rabbit Recombinant Polyclonal Antibody (Biotin)
77852-876
New Product
- Antigen Symbol:CDKN1A
- Clonality:Recombinant Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P38936
- Antigen Synonyms:CAP20|CDK-interacting protein 1|CDKI|CDKN1|CDN1A_HUMAN|CIP1|Cyclin Dependent Kinase Inhibitor 1A|Cyclin-dependent kinase inhibitor 1|Cyclin-dependent kinase inhibitor 1A (P21)|Cyclin-dependent kinase inhibitor 1A (p21, Cip1)|DNA Synthesis Inhibitor|MDA-6|MDA6|Melanoma differentiation-associated protein 6|Melanoma differentiation-associated protein|p21|P21 protein|p21CIP1|p21Cip1/Waf1|p21WAF|PIC1|SDI1|SLC12A9|WAF1|Wild type p53 activated fragment 1 (WAF1)|Wild type p53 activated fragment 1|Wildtype p53-activated fragment 1
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant human Cyclin-dependent kinase inhibitor 1 protein (2-164AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Type:
Antigen: CDKN1A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human