Anti-AXIN2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: AXIN2
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77846-319
- 77846-318
- Antigen Symbol:
- AXIN2
- AXIN2
- Antigen Synonyms:
- Axil|Axin like protein|Axin-2|Axin-like protein|Axin2|AXIN2_HUMAN|Axis inhibition protein 2|Conductin|DKFZp781B0869|MGC10366|MGC126582
- Axil|Axin like protein|Axin-2|Axin-like protein|Axin2|AXIN2_HUMAN|Axis inhibition protein 2|Conductin|DKFZp781B0869|MGC10366|MGC126582
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Peptide sequence from Human Axin-2 protein (56-73AA)
- Peptide sequence from Human Axin-2 protein (56-73AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together