Anti-ANAPC4 Rabbit Polyclonal Antibody
New Product
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:25 to 1:100
Type:
Antigen: ANAPC4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
- Catalog No:
- 77848-791
- 77848-790
- Antigen Symbol:
- ANAPC4
- ANAPC4
- Antigen Synonyms:
- ANAPC 4|anapc4|Anaphase Promoting Complex Subunit 4|Anaphase-promoting complex subunit 4|APC 4|APC Complex Protein CUT20|APC4|APC4_HUMAN|CUT20|Cyclosome subunit 4|LID1
- ANAPC 4|anapc4|Anaphase Promoting Complex Subunit 4|Anaphase-promoting complex subunit 4|APC 4|APC Complex Protein CUT20|APC4|APC4_HUMAN|CUT20|Cyclosome subunit 4|LID1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UJX5
- Q9UJX5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human ANAPC4
- Synthetic peptide of Human ANAPC4
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together