Anti-AMY2B Rabbit Recombinant Polyclonal Antibody
77851-128
New Product
- Antigen Symbol:AMY2B
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:P19961
- Antigen Synonyms:Alpha-amylase 2B|EC 3.2.1.1|1,4-alpha-D-glucan glucanohydrolase 2B|Carcinoid alpha-amylase
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Alpha-amylase 2B protein (16-320AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: WB: 1:200 to 1:1000, IHC: 1:20 to 1:200
Type:
Antigen: AMY2B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse