Anti-COL4A1 Rabbit Polyclonal Antibody
New Product
Recommended dilution: WB:1:1000 to 1:5000
Type:
Antigen: COL4A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
- Catalog No:
- 77849-241
- 77849-242
- Antigen Symbol:
- COL4A1
- COL4A1
- Antigen Synonyms:
- Arresten|BSVD|CO4A1_HUMAN|COL4A1|COL4A1 NC1 domain|COL4A2|COL4A3|COL4A4|COL4A5|collagen alpha-1(IV) chain|Collagen IV Alpha 1 Polypeptide|Collagen IV Alpha 2 Polypeptide|Collagen Of Basement Membrane Alpha 1 Chain|Collagen Of Basement Membrane Alpha 2 Chain|Collagen Type IV Alpha 1|collagen type IV alpha 1 chain|Collagen Type IV Alpha 2|Collagen Type IV Alpha 3|Collagen Type IV Alpha 4|Collagen Type IV Alpha 5|RATOR
- Arresten|BSVD|CO4A1_HUMAN|COL4A1|COL4A1 NC1 domain|COL4A2|COL4A3|COL4A4|COL4A5|collagen alpha-1(IV) chain|Collagen IV Alpha 1 Polypeptide|Collagen IV Alpha 2 Polypeptide|Collagen Of Basement Membrane Alpha 1 Chain|Collagen Of Basement Membrane Alpha 2 Chain|Collagen Type IV Alpha 1|collagen type IV alpha 1 chain|Collagen Type IV Alpha 2|Collagen Type IV Alpha 3|Collagen Type IV Alpha 4|Collagen Type IV Alpha 5|RATOR
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P02462
- P02462
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human COL4A1
- Synthetic peptide of Human COL4A1
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together