Anti-CTSE Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Catalog # 77853-763
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CTSE
- Clonality:Recombinant Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P14091
- Antigen Synonyms:CATE|CATE_HUMAN|Cathepsin E|Cathepsin E form II|Erythrocyte membrane aspartic proteinase|Slow moving proteinase
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Cathepsin E protein (59-155AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: CTSE
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human