Anti-BLOC1S1 Rabbit Recombinant polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 77852-747
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:BLOC1S1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P78537
- Antigen Synonyms:BLOS1|GCN5L1|RT14Biogenesis of lysosome-related organelles complex 1 subunit 1|BLOC-1 subunit 1|GCN5-like protein 1|Protein RT14
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Shipping Temperature:Shipped on Blue ice
- Immunogen:Recombinant Human Biogenesis of lysosome-related organelles complex 1 subunit 1 protein (1-153AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
0
Type:
Antigen: BLOC1S1
Clonality: Recombinant polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human