Anti-CSNK1G1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product
Type:
Antigen: CSNK1G1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77843-296
- 77843-297
- Antigen Symbol:
- CSNK1G1
- CSNK1G1
- Antigen Synonyms:
- 9130020E21Rik|Casein kinase 1 gamma 1|Casein kinase I isoform gamma 1|Casein kinase I isoform gamma-1|CKI gamma 1|CKI-gamma 1|EC 2.7.11.1|KC1G1_HUMAN
- 9130020E21Rik|Casein kinase 1 gamma 1|Casein kinase I isoform gamma 1|Casein kinase I isoform gamma-1|CKI gamma 1|CKI-gamma 1|EC 2.7.11.1|KC1G1_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9HCP0
- Q9HCP0
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Casein kinase I isoform gamma-1 protein (1-125AA)
- Recombinant Human Casein kinase I isoform gamma-1 protein (1-125AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together