Anti-CRHR2 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB:1:1000 to 1:5000
Type:
Antigen: CRHR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77849-391
- 77849-390
- Antigen Symbol:
- CRHR2
- CRHR2
- Antigen Synonyms:
- Corticotropin releasing hormone receptor 2|Corticotropin-releasing factor receptor 2|Corticotropin-releasing hormone receptor 2|CRF 2|CRF 2R|CRF R2|CRF RB|CRF-R-2|CRF-R2|CRF2|CRF2 receptor beta isoform|CRF2R|CRFR 2|CRFR-2|CRFR2|CRFR2_HUMAN|CRH 2R|CRH R2|CRH-R-2|CRH-R2|CRH2R|CRHR 2|Crhr2|HM CRF
- Corticotropin releasing hormone receptor 2|Corticotropin-releasing factor receptor 2|Corticotropin-releasing hormone receptor 2|CRF 2|CRF 2R|CRF R2|CRF RB|CRF-R-2|CRF-R2|CRF2|CRF2 receptor beta isoform|CRF2R|CRFR 2|CRFR-2|CRFR2|CRFR2_HUMAN|CRH 2R|CRH R2|CRH-R-2|CRH-R2|CRH2R|CRHR 2|Crhr2|HM CRF
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q13324
- Q13324
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human CRHR2
- Synthetic peptide of Human CRHR2
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together