Anti-CLASP2 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
77853-177
New Product
- Antigen Symbol:CLASP2
- Clonality:Recombinant Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Form:Liquid
- Gene ID:O75122
- Antigen Synonyms:CLAP2_HUMAN|CLASP 2|Clasp2|CLIP associating protein 2|CLIP-associating protein 2|Cytoplasmic linker associated protein 2|Cytoplasmic linker-associated protein 2|hOrbit2|KIAA0627|Multiple asters (Mast) like homolog 2|Protein Orbit homolog 2
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human CLIP-associating protein 2 protein (226-381AA)
- Purification:>95% Protein G purified
- Packaging:Vial
Type:
Antigen: CLASP2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human