Anti-HTRA2 Rabbit Recombinant Polyclonal Antibody
Catalog # 77857-765
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:HTRA2
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:O43464
- Antigen Synonyms:High temperature requirement protein A2|HTRA 2|HtrA like serine protease|HtrA serine peptidase 2|HtrA|E. coli|homolog of|2|HtrA2|HTRA2_HUMAN|mitochondrial|Omi stress regulated endoprotease|Omi stress-regulated endoprotease|PARK 13|PARK13|Protease serine 25|PRSS 25|PRSS25|Serine protease 25|Serine protease HTRA2|Serine protease HTRA2 mitochondrial|Serine protease htra2 mitochondrial precursor|Serine protease omi|Serine proteinase OMI
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Serine protease HTRA2, mitochondrial protein (334-458AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:20 to 1:200, IP: 1:200 to 1:2000
Type:
Antigen: HTRA2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human