Anti-Chrm1 Rabbit Recombinant Polyclonal Antibody
77853-098
New Product
- Antigen Symbol:Chrm1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rat
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P08482
- Antigen Synonyms:Chrm-1Muscarinic acetylcholine receptor M1
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Rat Muscarinic acetylcholine receptor M1 protein (210-366AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:5000
Type:
Antigen: Chrm1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat