Anti-PIH1D1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PIH1D1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77843-574
- 77843-573
- Antigen Symbol:
- PIH1D1
- PIH1D1
- Antigen Synonyms:
- NOP17|Nucleolar protein 17 homolog|PIH1 domain containing 1|PIH1 domain containing protein 1|PIH1 domain-containing protein 1|PIH1D1|PIHD1_HUMAN
- NOP17|Nucleolar protein 17 homolog|PIH1 domain containing 1|PIH1 domain containing protein 1|PIH1 domain-containing protein 1|PIH1D1|PIHD1_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NWS0
- Q9NWS0
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human PIH1 domain-containing protein 1 protein (1-290AA)
- Recombinant Human PIH1 domain-containing protein 1 protein (1-290AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together