Anti-SLC7A7 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SLC7A7
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77844-980
- 77844-979
- Antigen Symbol:
- SLC7A7
- SLC7A7
- Antigen Synonyms:
- LAT3|LPI|Monocyte amino acid permease 2|MOP-2|MOP2|SLC7A 7|Slc7a7|Solute carrier family 7 (cationic amino acid transporter|y+ system)|member 7|Solute carrier family 7 member 7|y(+)L type amino acid transporter 1|y(+)L-type amino acid transporter 1|Y+L amino acid transporter 1|y+LAT-1|Y+LAT1|YLAT1_HUMAN
- LAT3|LPI|Monocyte amino acid permease 2|MOP-2|MOP2|SLC7A 7|Slc7a7|Solute carrier family 7 (cationic amino acid transporter|y+ system)|member 7|Solute carrier family 7 member 7|y(+)L type amino acid transporter 1|y(+)L-type amino acid transporter 1|Y+L amino acid transporter 1|y+LAT-1|Y+LAT1|YLAT1_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UM01
- Q9UM01
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Y+L amino acid transporter 1 protein (325-382AA)
- Recombinant Human Y+L amino acid transporter 1 protein (325-382AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together