Anti-MACF1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77844-145
- 77844-144
- Antigen Symbol:
- MACF1
- MACF1
- Antigen Synonyms:
- MACF1|ABP620|ACF7|KIAA0465|KIAA1251|Microtubule-actin cross-linking factor 1|isoforms 1/2/3/5|620 kDa actin-binding protein|Actin cross-linking family protein 7|Macrophin-1|Trabeculin-alpha
- MACF1|ABP620|ACF7|KIAA0465|KIAA1251|Microtubule-actin cross-linking factor 1|isoforms 1/2/3/5|620 kDa actin-binding protein|Actin cross-linking family protein 7|Macrophin-1|Trabeculin-alpha
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UPN3
- Q9UPN3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Microtubule-actin cross-linking factor 1, isoforms 1/2/3/5 protein (1936-2150AA)
- Recombinant Human Microtubule-actin cross-linking factor 1, isoforms 1/2/3/5 protein (1936-2150AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together