Anti-CD151 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Catalog # 77852-608
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CD151
- Clonality:Recombinant Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P48509
- Antigen Synonyms:CD151 antigen|CD151 molecule (Raph blood group)|CD151 molecule|CD151_HUMAN|GP27|hemidesmosomal tetraspanin Membrane glycoprotein SFA-1|MER2|PETA-3|PETA3|Platelet endothelial tetraspan antigen 3|platelet surface glycoprotein gp27|Platelet-endothelial cell tetraspan antigen 3|Platelet-endothelial tetraspan antigen 3|RAPH|Red blood cell antigen MER2|SFA 1|SFA1|Tetraspanin-24|Tspan-24|TSPAN24
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human CD151 antigen protein (125-199AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: CD151
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human