Anti-CDH16 Rabbit Recombinant Polyclonal Antibody
Catalog # 77852-832
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CDH16
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Form:Liquid
- Gene ID:O75309
- Antigen Synonyms:CAD16_HUMAN|Cadherin 16|Cadherin 16 KSP cadherin|Cadherin kidney|Cadherin-16|CDH 16|CDH16|Kidney cadherin|Kidney specific cadherin|Kidney-specific cadherin|Ksp cadherin |Ksp-cadherin|PRO1340|UNQ695
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Cadherin-16 protein (19-230AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC:1:20 to 1:200
Type:
Antigen: CDH16
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human