Anti-ABCC9 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 77850-538
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ABCC9
- Clonality:Recombinant Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:O60706
- Antigen Synonyms:ABC37|ABCC9_HUMAN|AI414027|AI449286|ATFB12|ATP-binding cassette sub-family C member 9|ATP-binding cassette transporter sub-family C member 9|ATP-binding cassette|sub-family C (CFTR/MRP)|member 9|CANTU|CMD1O|FLJ36852|Sulfonylurea receptor 2|Sulfonylurea-binding protein 2|SUR2|SUR2A|SUR2B
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human ATP-binding cassette sub-family C member 9 protein (560-669AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human