Anti-APOA2 HEK293F cells Recombinant Monoclonal Antibody [clone: 6H2]
Catalog # 77850-154
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:APOA2
- Clonality:Recombinant Monoclonal
- Clone:6H2
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:HEK293F cells
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:P02652
- Antigen Synonyms:Apolipoprotein A-II, Apo-AII, ApoA-II, Apolipoprotein A2, Proapolipoprotein A-II, ProapoA-II, Truncated apolipoprotein A-II, Apolipoprotein A-II(1-76), APOA2
- Storage Buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:A synthesized peptide derived from Human APOA2
- Purification:Affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:50 to 1:200
Type:
Antigen: APOA2
Clonality: Recombinant Monoclonal
Clone: 6H2
Conjugation: Unconjugated
Epitope:
Host: HEK293F cells
Isotype: IgG
Reactivity: Human