Anti-CHRM3 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Catalog # 77853-111
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CHRM3
- Clonality:Recombinant Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P20309
- Antigen Synonyms:AChR|ACM3_HUMAN|cholinergic receptor muscarinic 3|CHRM 3|EGBRS|HM 3|HM3|m3 muscarinic acetylcholine receptor|M3 muscarinic receptor|muscarinic 3|Muscarinic acetylcholine receptor M3|muscarinic cholinergic m3 receptor|muscarinic m3 receptor
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant human Muscarinic acetylcholine receptor M3 protein (253-492AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: CHRM3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human