Anti-SPO11 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SPO11
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77846-419
- 77846-420
- Antigen Symbol:
- SPO11
- SPO11
- Antigen Synonyms:
- Cancer/testis antigen 35|CT35|Meiotic recombination protein SPO 11|Meiotic recombination protein SPO11|MGC39953|SPATA43|Spermatogenesis associated 43|SPO 11|SPO11|SPO11 meiotic protein covalently bound to DSB|SPO11 meiotic protein covalently bound to DSB homolog|SPO11 meiotic protein covalently bound to DSB like|SPO11_HUMAN|TOPVIA
- Cancer/testis antigen 35|CT35|Meiotic recombination protein SPO 11|Meiotic recombination protein SPO11|MGC39953|SPATA43|Spermatogenesis associated 43|SPO 11|SPO11|SPO11 meiotic protein covalently bound to DSB|SPO11 meiotic protein covalently bound to DSB homolog|SPO11 meiotic protein covalently bound to DSB like|SPO11_HUMAN|TOPVIA
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y5K1
- Q9Y5K1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Meiotic recombination protein SPO11 protein (28-109AA)
- Recombinant Human Meiotic recombination protein SPO11 protein (28-109AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95% Protein G purified
- >95% Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together