Anti-CHUK Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:2000, WB: 1:200 to 1:1000, IHC: 1:25 to 1:100
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77849-315
- 77849-316
- Antigen Symbol:
- CHUK
- CHUK
- Antigen Synonyms:
- chuk|CHUK1|Conserved Helix Loop Helix Ubiquitous Kinase|Conserved helix loop ubiquitous kinase|Conserved helix-loop-helix ubiquitous kinase|I Kappa B Kinase 1|I Kappa B Kinase Alpha|I-kappa-B kinase 1|I-kappa-B kinase alpha|IkappaB kinase|IkB kinase alpha subunit|IkBKA|IKK 1|IKK A|IKK a kinase|IKK-A|IKK-alpha|IKK1|IKKA|IKKA_HUMAN|Inhibitor Of Kappa Light Polypeptide Gene Enhancer In B Cells|Inhibitor Of Nuclear Factor Kappa B Kinase Alpha Subunit|Inhibitor of nuclear factor kappa-B kinase subunit alpha|NFKBIKA|Nuclear Factor Kappa B Inhibitor Kinase Alpha|Nuclear factor NF kappa B inhibitor kinase alpha|Nuclear factor NF-kappa-B inhibitor kinase alpha|Nuclear factor NFkappaB inhibitor kinase alpha|Nuclear Factor Of Kappa Light Chain Gene Enhancer In B Cells Inhibitor|TCF-16|TCF16|Transcription factor 16
- chuk|CHUK1|Conserved Helix Loop Helix Ubiquitous Kinase|Conserved helix loop ubiquitous kinase|Conserved helix-loop-helix ubiquitous kinase|I Kappa B Kinase 1|I Kappa B Kinase Alpha|I-kappa-B kinase 1|I-kappa-B kinase alpha|IkappaB kinase|IkB kinase alpha subunit|IkBKA|IKK 1|IKK A|IKK a kinase|IKK-A|IKK-alpha|IKK1|IKKA|IKKA_HUMAN|Inhibitor Of Kappa Light Polypeptide Gene Enhancer In B Cells|Inhibitor Of Nuclear Factor Kappa B Kinase Alpha Subunit|Inhibitor of nuclear factor kappa-B kinase subunit alpha|NFKBIKA|Nuclear Factor Kappa B Inhibitor Kinase Alpha|Nuclear factor NF kappa B inhibitor kinase alpha|Nuclear factor NF-kappa-B inhibitor kinase alpha|Nuclear factor NFkappaB inhibitor kinase alpha|Nuclear Factor Of Kappa Light Chain Gene Enhancer In B Cells Inhibitor|TCF-16|TCF16|Transcription factor 16
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O15111
- O15111
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human CHUK
- Synthetic peptide of Human CHUK
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together