Anti-DNAJB2 Rabbit Recombinant Polyclonal Antibody
Catalog # 77854-250
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:DNAJB2
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P25686
- Antigen Synonyms:HSJ1|HSPF3DnaJ homolog subfamily B member 2|Heat shock 40 kDa protein 3|Heat shock protein J1|HSJ-1
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human DnaJ homolog subfamily B member 2 protein (2-324AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:20 to 1:200
Type:
Antigen: DNAJB2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human