Anti-ANGPTL5 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:20 to 1:100
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77849-291
- 77849-290
- Antigen Symbol:
- ANGPTL5
- ANGPTL5
- Antigen Synonyms:
- Angiopoietin like 5|Angiopoietin related protein 5|Angiopoietin-like protein 5|Angiopoietin-related protein 5|ANGL5_HUMAN|ANGPTL 5|ANGPTL5|Fibrinogen like
- Angiopoietin like 5|Angiopoietin related protein 5|Angiopoietin-like protein 5|Angiopoietin-related protein 5|ANGL5_HUMAN|ANGPTL 5|ANGPTL5|Fibrinogen like
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q86XS5
- Q86XS5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human ANGPTL5
- Synthetic peptide of Human ANGPTL5
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together