Anti-MLYCD Rabbit Recombinant Polyclonal Antibody
Catalog # 77859-833
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:MLYCD
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:O95822
- Antigen Synonyms:DCMC_HUMAN|hMCD|Malonyl CoA decarboxylase|Malonyl CoA decarboxylase mitochondrial|Malonyl coenzyme A decarboxylase|Malonyl-CoA decarboxylase|MCD|MGC59795|mitochondrial|Mlycd
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Malonyl-CoA decarboxylase, mitochondrial protein (40-493AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:2000 to 1:5000, IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: MLYCD
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human