Anti-ARNTL Rabbit Recombinant Polyclonal Antibody
Catalog # 77851-406
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ARNTL
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:O00327
- Antigen Synonyms:ARNT like protein 1 brain and muscle|Aryl hydrocarbon receptor nuclear translocator like|Aryl hydrocarbon receptor nuclear translocator like protein 1|Aryl hydrocarbon receptor nuclear translocator-like protein 1|Basic helix loop helix PAS orphan MOP3|Basic helix loop helix PAS protein MOP3|Basic-helix-loop-helix-PAS protein MOP3|bHLH PAS protein JAP3|bHLH-PAS protein JAP3|bHLHe5|BMAL 1|BMAL1_HUMAN|BMAL1c|Brain and muscle ARNT like 1|Brain and muscle ARNT-like 1|CG8727 PA|Class E basic helix-loop-helix protein 5|cycle|JAP 3|JAP3|Member of PAS protein 3|Member of PAS superfamily 3|MGC47515|MOP 3|MOP3|PAS domain-containing protein 3|PASD 3|PASD3|TIC
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant human Aryl hydrocarbon receptor nuclear translocator-like protein 1 protein (366-585AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IF: 1:50 to 1:200
Type:
Antigen: ARNTL
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human