Anti-BNIP1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:200 to 1:1000, IHC: 1:50 to 1:200
Type:
Antigen: BNIP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77848-728
- 77848-727
- Antigen Symbol:
- BNIP1
- BNIP1
- Antigen Synonyms:
- BCL2 adenovirus E1B 19kD interacting protein 1|BCL2/adenovirus E1B 19 kDa protein-interacting protein 1|BNIP1|MGC41600|Nip1|OTTHUMP00000161077|OTTHUMP00000161078|OTTHUMP00000161079|OTTHUMP00000223580|sec20|SEC20_HUMAN|SEC20L|Transformation-related gene 8 protein|TRG-8|TRG8|Vesicle transport protein SEC20
- BCL2 adenovirus E1B 19kD interacting protein 1|BCL2/adenovirus E1B 19 kDa protein-interacting protein 1|BNIP1|MGC41600|Nip1|OTTHUMP00000161077|OTTHUMP00000161078|OTTHUMP00000161079|OTTHUMP00000223580|sec20|SEC20_HUMAN|SEC20L|Transformation-related gene 8 protein|TRG-8|TRG8|Vesicle transport protein SEC20
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q12981
- Q12981
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human BNIP1
- Fusion protein of Human BNIP1
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together