Anti-CHRNB2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Catalog # 77853-129
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CHRNB2
- Clonality:Recombinant Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P17787
- Antigen Synonyms:Acetylcholine receptor beta 2 neural|ACHB2_HUMAN|ACHN|AChR|Acrb 2|Acrb2|b2 nAchR|Cholinergic receptor nicotinic beta 2|Cholinergic receptor nicotinic beta polypeptide 2|Cholinergic receptor nicotinic beta polypeptide 2 neuronal|cholinergic receptor|nicotinic|beta 2 (neuronal)|Chrnb2|EFNL 3|EFNL3|nAChRB2|Neuronal acetylcholine receptor protein beta 2 chain precursor|Neuronal acetylcholine receptor protein subunit beta 2|Neuronal acetylcholine receptor subunit beta-2|Neuronal nicotinic acetylcholine receptor beta 2
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Neuronal acetylcholine receptor subunit beta-2 protein (322-458AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: CHRNB2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human