Anti-SALL4 Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: SALL4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77844-723
- 77844-722
- Antigen Symbol:
- SALL4
- SALL4
- Antigen Synonyms:
- AA407717|AL022809|AW536104|C330011P20Rik|C78083|C78563|dJ1112F19.1|DRRS|HSAL4|Sal like 4 (Drosophila)|Sal like 4|Sal like Protein 4|Sal-like protein 4|Sall4|SALL4_HUMAN|Spalt like transcription factor 4|Tex20|Zinc finger protein 797|Zinc finger protein SALL4|ZNF797
- AA407717|AL022809|AW536104|C330011P20Rik|C78083|C78563|dJ1112F19.1|DRRS|HSAL4|Sal like 4 (Drosophila)|Sal like 4|Sal like Protein 4|Sal-like protein 4|Sall4|SALL4_HUMAN|Spalt like transcription factor 4|Tex20|Zinc finger protein 797|Zinc finger protein SALL4|ZNF797
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UJQ4
- Q9UJQ4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Sal-like protein 4 protein (1-220AA)
- Recombinant Human Sal-like protein 4 protein (1-220AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together