Anti-CYP21A2 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:200 to 1:1000, IHC: 1:50 to 1:200
Type:
Antigen: CYP21A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77844-155
- 77844-154
- Antigen Symbol:
- CYP21A2
- CYP21A2
- Antigen Synonyms:
- CYP21|21 OHase|21-OHase|CA21H|CAH1|CP21A_HUMAN|CPS1|CYP21A 2|CYP21B|Cytochrome P-450c21|Cytochrome P450 21|Cytochrome P450 C21B|Cytochrome P450 XXI|Cytochrome P450|family 21|subfamily A|polypeptide 2|subfamily XXIA (steroid 21 hydroxylase|congenital adrenal hyperplasia)|Cytochrome P450-C21|Cytochrome P450-C21B|P450 C21|P450 C21B|P450c21B|Steroid 21 hydroxylase|Steroid 21 monooxygenase|Steroid 21-hydroxylase
- CYP21|21 OHase|21-OHase|CA21H|CAH1|CP21A_HUMAN|CPS1|CYP21A 2|CYP21B|Cytochrome P-450c21|Cytochrome P450 21|Cytochrome P450 C21B|Cytochrome P450 XXI|Cytochrome P450|family 21|subfamily A|polypeptide 2|subfamily XXIA (steroid 21 hydroxylase|congenital adrenal hyperplasia)|Cytochrome P450-C21|Cytochrome P450-C21B|P450 C21|P450 C21B|P450c21B|Steroid 21 hydroxylase|Steroid 21 monooxygenase|Steroid 21-hydroxylase
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P08686
- P08686
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human CYP21A2
- Fusion protein of Human CYP21A2
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together