Anti-YARS2 Rabbit Recombinant polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB:1:500-1:2000, IHC:1:20-1:200
Type:
Antigen: YARS2
Clonality: Recombinant polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77845-868
- 77845-869
- Antigen Symbol:
- YARS2
- YARS2
- Antigen Synonyms:
- CGI 04|FLJ13995|mitochondrial|MLASA2|mt TyrRS|SYYM_HUMAN|Tyrosine tRNA ligase 2 mitochondrial|Tyrosine tRNA ligase|Tyrosine tRNA ligase mitochondrial|Tyrosine--tRNA ligase|Tyrosyl tRNA synthetase 2 mitochondrial|Tyrosyl tRNA synthetase mitochondrial precursor|Tyrosyl-tRNA synthetase|TyrRS|YARS 2|YARS2
- CGI 04|FLJ13995|mitochondrial|MLASA2|mt TyrRS|SYYM_HUMAN|Tyrosine tRNA ligase 2 mitochondrial|Tyrosine tRNA ligase|Tyrosine tRNA ligase mitochondrial|Tyrosine--tRNA ligase|Tyrosyl tRNA synthetase 2 mitochondrial|Tyrosyl tRNA synthetase mitochondrial precursor|Tyrosyl-tRNA synthetase|TyrRS|YARS 2|YARS2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y2Z4
- Q9Y2Z4
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Tyrosine--tRNA ligase, mitochondrial protein (228-477AA)
- Recombinant Human Tyrosine--tRNA ligase, mitochondrial protein (228-477AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together