Anti-FHOD1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: FHOD1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77845-488
- 77845-489
- Antigen Symbol:
- FHOD1
- FHOD1
- Antigen Synonyms:
- FH1/FH2 domain containing protein|FH1/FH2 domain containing protein 1|FH1/FH2 domain-containing protein 1|FHOD1_HUMAN|FHOS|FHOS1|Formin homolog overexpressed in spleen 1|Formin homology 2 domain containing 1|Formin homology 2 domain containing protein 1|Formin homology 2 domain-containing protein 1
- FH1/FH2 domain containing protein|FH1/FH2 domain containing protein 1|FH1/FH2 domain-containing protein 1|FHOD1_HUMAN|FHOS|FHOS1|Formin homolog overexpressed in spleen 1|Formin homology 2 domain containing 1|Formin homology 2 domain containing protein 1|Formin homology 2 domain-containing protein 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y613
- Q9Y613
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant human FH1/FH2 domain-containing protein 1 protein (338-437AA)
- Recombinant human FH1/FH2 domain-containing protein 1 protein (338-437AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together