Anti-SUMO1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:2000, IHC: 1:15 to 1:50
Type:
Antigen: SUMO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- 77847-226
- 77847-225
- Antigen Symbol:
- SUMO1
- SUMO1
- Antigen Synonyms:
- DAP1|GAP modifying protein 1|GAP-modifying protein 1|GMP 1|GMP1|OFC10|PIC 1|PIC1|SENP2|Sentrin 1|Sentrin|Small ubiquitin related modifier 1|Small ubiquitin-like modifier 1|Small ubiquitin-related modifier 1|SMT3|SMT3 homolog 3|SMT3 suppressor of mif two 3 homolog 1|SMT3, yeast, homolog 3|Smt3C|SMT3H3|SUMO-1|SUMO1_HUMAN|Ubiquitin homology domain protein PIC1|Ubiquitin Like 1|Ubiquitin like protein SMT3C|Ubiquitin like protein UBL1|Ubiquitin-homology domain protein PIC1|Ubiquitin-like protein SMT3C|Ubiquitin-like protein UBL1|UBL 1|UBL1
- DAP1|GAP modifying protein 1|GAP-modifying protein 1|GMP 1|GMP1|OFC10|PIC 1|PIC1|SENP2|Sentrin 1|Sentrin|Small ubiquitin related modifier 1|Small ubiquitin-like modifier 1|Small ubiquitin-related modifier 1|SMT3|SMT3 homolog 3|SMT3 suppressor of mif two 3 homolog 1|SMT3, yeast, homolog 3|Smt3C|SMT3H3|SUMO-1|SUMO1_HUMAN|Ubiquitin homology domain protein PIC1|Ubiquitin Like 1|Ubiquitin like protein SMT3C|Ubiquitin like protein UBL1|Ubiquitin-homology domain protein PIC1|Ubiquitin-like protein SMT3C|Ubiquitin-like protein UBL1|UBL 1|UBL1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human SUMO1
- Synthetic peptide of Human SUMO1
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together