Anti-Snaclec rhodocytin subunit alpha Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: Snaclec rhodocytin subunit alpha
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Calloselasma rhodostoma
- Catalog No:
- 77843-311
- 77843-310
- Antigen Symbol:
- Snaclec rhodocytin subunit alpha
- Snaclec rhodocytin subunit alpha
- Antigen Synonyms:
- antibody|Aggretin alpha chain|Rhodoaggretin subunit alpha
- antibody|Aggretin alpha chain|Rhodoaggretin subunit alpha
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Calloselasma rhodostoma
- Calloselasma rhodostoma
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9I841
- Q9I841
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Calloselasma rhodostoma Snaclec rhodocytin subunit alpha protein (1-136AA)
- Recombinant Calloselasma rhodostoma Snaclec rhodocytin subunit alpha protein (1-136AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together