Anti-DOK5 Rabbit Recombinant Polyclonal Antibody
77854-318
New Product
- Antigen Symbol:DOK5
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q9P104
- Antigen Synonyms:chromosome 20 open reading frame 180|Docking protein 5|DOK 5|DOK5 protein|DOK5_HUMAN|Downstream of tyrosine kinase 5|Insulin receptor substrate 6|IRS-6|IRS6
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Docking protein 5 protein (1-198AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Recommended dilution: WB:1:1000 to 1:5000, IHC:1:20 to 1:200, IF:1:50 to 1:200
Type:
Antigen: DOK5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse