Anti-CD300LB Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # 77852-645
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CD300LB
- Clonality:Recombinant Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:A8K4G0
- Antigen Synonyms:CD300B|CLM7|CMRF35A2|IREM3|LMIR5|TREM5|UNQ2530/PRO6029CMRF35-like molecule 7|CLM-7|CD300 antigen-like family member B|CMRF35-A2|Immune receptor expressed on myeloid cells 3|IREM-3|Leukocyte mono-Ig-like receptor 5|Triggering receptor expressed on myeloid cells 5|TREM-5|CD antigen CD300b
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human CMRF35-like molecule 7 protein (18-151AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: CD300LB
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human