Anti-RRAGC Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:25 to 1:100
Type:
Antigen: RRAGC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Specifications
- Catalog No:
- 77847-877
- 77847-878
- Antigen Symbol:
- RRAGC
- RRAGC
- Antigen Synonyms:
- AU041672|FLJ13311|GTPase interacting protein 2|GTPase-interacting protein 2|GTR2|MGC47404|OTTHUMP00000000548|Rag C|Rag C protein|RAGC|Ras related GTP binding C|Ras-related GTP-binding protein C|RP23 29H22.4|RRAGC_HUMAN|TIB929|YGR163W
- AU041672|FLJ13311|GTPase interacting protein 2|GTPase-interacting protein 2|GTR2|MGC47404|OTTHUMP00000000548|Rag C|Rag C protein|RAGC|Ras related GTP binding C|Ras-related GTP-binding protein C|RP23 29H22.4|RRAGC_HUMAN|TIB929|YGR163W
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9HB90
- Q9HB90
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human RRAGC
- Fusion protein of Human RRAGC
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together