Anti-CMA1 Rabbit Recombinant Polyclonal Antibody
77853-289
New Product
- Antigen Symbol:CMA1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µl
- Form:Liquid
- Gene ID:P23946
- Antigen Synonyms:Alpha-chymase|Chymase 1|Chymase 1 mast cell|chymase 1 preproprotein transcript E|chymase 1 preproprotein transcript I|Chymase|heart|mast cell|CMA1_HUMAN|CYH|CYM|EC 3.4.21.39|Mast cell chymase 1|Mast cell protease 3|Mast cell protease 5|Mast cell protease I|Mast cell protease III|Mcp-5|MCP3P|Mcpt5|MCT1|MGC119890|MGC119891|MMCP-5
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Chymase protein (1-247AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: CMA1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human