Anti-SSR3 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB:1:1000 to 1:5000
Type:
Antigen: SSR3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77845-016
- 77845-017
- Antigen Symbol:
- SSR3
- SSR3
- Antigen Synonyms:
- Signal sequence receptor gamma| Signal sequence receptor subunit gamma| SSR-gamma| Ssr3| SSRG_HUMAN| Translocon associated protein gamma subunit| Translocon-associated protein subunit gamma| TRAP complex gamma subuni| TRAP-gamma| TRAPG
- Signal sequence receptor gamma| Signal sequence receptor subunit gamma| SSR-gamma| Ssr3| SSRG_HUMAN| Translocon associated protein gamma subunit| Translocon-associated protein subunit gamma| TRAP complex gamma subuni| TRAP-gamma| TRAPG
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UNL2
- Q9UNL2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Translocon-associated protein subunit gamma protein (77-135AA)
- Recombinant Human Translocon-associated protein subunit gamma protein (77-135AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together