Anti-CCT6A Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 77852-598
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CCT6A
- Clonality:Recombinant Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P40227
- Antigen Synonyms:CCT6|CCTZ|T-complex protein 1 subunit zeta|TCP-1-zeta|Acute morphine dependence-related protein 2|CCT-zeta-1|HTR3|Tcp20
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human T-complex protein 1 subunit zeta protein (80-250AA)
- Purification:>95%, protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: CCT6A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human