Anti-FBXO28 Rabbit Recombinant polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: FBXO28
Clonality: Recombinant polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77843-531
- 77843-530
- Antigen Symbol:
- FBXO28
- FBXO28
- Antigen Synonyms:
- CENP 30|Centromere protein 30|F box only protein 28|F box protein 28|F-box only protein 28|Fbx28 |FBX28_HUMAN|FLJ10766|KIAA0483|OTTHUMP00000035597
- CENP 30|Centromere protein 30|F box only protein 28|F box protein 28|F-box only protein 28|Fbx28 |FBX28_HUMAN|FLJ10766|KIAA0483|OTTHUMP00000035597
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NVF7
- Q9NVF7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human F-box only protein 28 protein (251-368AA)
- Recombinant Human F-box only protein 28 protein (251-368AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together