Anti-ARRB2 Rabbit Recombinant Polyclonal Antibody
Catalog # 77851-422
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ARRB2
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western Blot:Yes
- Form:Liquid
- Gene ID:P32121
- Antigen Synonyms:ARB 2|ARB2|ARR 2|ARR2|ARRB 2|ARRB2|ARRB2_HUMAN|Arrestin 3|Arrestin beta 2|Arrestin beta-2|BARR2|Beta-arrestin-2|DKFZp686L0365|HGNC:712
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Beta-arrestin-2 protein (222-369AA)
- Purification:>95% Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:500 to 1:5000, IHC:1:500 to 1:1000, IF:1:50 to 1:200
Type:
Antigen: ARRB2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat