Anti-NIP7 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: NIP7
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77846-048
- 77846-049
- Antigen Symbol:
- NIP7
- NIP7
- Antigen Synonyms:
- 60S ribosome subunit biogenesis protein NIP7 homolog|CGI 37|CGI37|FLJ10296|HSPC031|HSPC180|KD93|NIP 7|NIP7|NIP7 nucleolar pre rRNA processing protein|NIP7_HUMAN|Nuclear import 7|Nuclear import 7 homolog (S. cerevisiae)|Nuclear import 7 homolog|OK/SW cl.76|OK/SW cl.78|OTTHUMP00000174889
- 60S ribosome subunit biogenesis protein NIP7 homolog|CGI 37|CGI37|FLJ10296|HSPC031|HSPC180|KD93|NIP 7|NIP7|NIP7 nucleolar pre rRNA processing protein|NIP7_HUMAN|Nuclear import 7|Nuclear import 7 homolog (S. cerevisiae)|Nuclear import 7 homolog|OK/SW cl.76|OK/SW cl.78|OTTHUMP00000174889
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y221
- Q9Y221
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Recombinant Human 60S ribosome subunit biogenesis protein NIP7 homolog protein (1-180AA)
- Recombinant Human 60S ribosome subunit biogenesis protein NIP7 homolog protein (1-180AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together