Anti-DYSF Rabbit Recombinant Polyclonal Antibody
Catalog # 77854-449
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:DYSF
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:O75923
- Antigen Synonyms:DMAT|DYSF|DYSF_HUMAN|Dysferlin|Dysferlin limb girdle muscular dystrophy 2B (autosomal recessive)|Dysferlin limb girdle muscular dystrophy 2B|Dystrophy associated fer 1 like 1|Dystrophy associated fer 1 like protein|Dystrophy associated fer1 like 1|Dystrophy associated fer1 like protein|Dystrophy-associated fer-1-like protein|Fer 1 like protein 1|Fer-1-like protein 1|Fer1 like protein 1|FER1L1|FLJ00175|FLJ90168|LGMD 2B|LGMD2B|Limb girdle muscular dystrophy 2B (autosomal recessive)|Limb girdle muscular dystrophy 2B|Miyoshi myopathy|MM|MMD1
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Dysferlin protein (105-278AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: DYSF
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human