Anti-ADAP2 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ADAP2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77843-361
- 77843-362
- Antigen Symbol:
- ADAP2
- ADAP2
- Antigen Synonyms:
- Adap2|ADAP2_HUMAN|Arf-GAP with dual PH domain-containing protein 2|ArfGAP with dual PH domains 2|cent b|Centa 2|CENTA2|Centaurin alpha 2|Centaurin alpha 2 protein|Centaurin beta|Centaurin-alpha-2|Cnt-a2|HSA272195
- Adap2|ADAP2_HUMAN|Arf-GAP with dual PH domain-containing protein 2|ArfGAP with dual PH domains 2|cent b|Centa 2|CENTA2|Centaurin alpha 2|Centaurin alpha 2 protein|Centaurin beta|Centaurin-alpha-2|Cnt-a2|HSA272195
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Arf-GAP with dual PH domain-containing protein 2 protein (110-381AA)
- Recombinant Human Arf-GAP with dual PH domain-containing protein 2 protein (110-381AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together